missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MUC4 (aa 4972-5104) Control Fragment Recombinant Protein

Product Code. 30203165
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203165

Brand: Invitrogen™ RP88564

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (62%), Rat (62%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Nearly all mucosal surfaces are protected by mucin transmembrane glycoproteins, from bronchioles to corneas to reproductive tracts. Mucin 4 (MUC4) is membrane-bound and is important in protecting and lubricating. MUC4 expression is erroneous in many carcinomas and sarcomas, including fibromyoxoid sarcoma and pancreatic adenocarcinoma. Immunohistochemical staining for MUC4 has been discussed as a potential biomarker for detection of these cancers. MUC4 seems to be most predominantly affected in Asian diseases - Taiwanese infertility and endometriosis, as well as gallstone formation in Chinese men, are connected to MUC4, where similar diseases in other populations do not share these MUC4 mutations. Because MUC4 is so important in mucosal surface protection, it is not surprising that it has been implicated as failing in many metastatic cancer invasions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99102
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4585
Name Human MUC4 (aa 4972-5104) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4933405I11Rik; Ascites sialoglycoprotein; Ascites sialoglycoprotein 1; Ascites sialoglycoprotein 2; ASGP; ASGP1; ASGP-1; ASGP-2; HSA276359; MUC4; MUC-4; mucin 4; mucin 4, ASGP; mucin 4, cell surface associated; mucin 4, tracheobronchial; Mucin4; mucin-4; Mucin-4 alpha chain; Mucin-4 beta chain; Pancreatic adenocarcinoma mucin; pre-sialomucin complex; Psmc; sialomucin ascites sialoglycoprotein-1; sialomucin complex; testis mucin; Tracheobronchial mucin
Common Name MUC4
Gene Symbol MUC4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IQYTSNAEDANFTLRDSCTDLELFENGTLLWTPKSLEPFTLEILARSAKIGLASALQPRTVVCHCNAESQCLYNQTSRVGNSSLEVAGCKCDGGTFGRYCEGSEDACEEPCFPSVHCVPGKGCEACPPNLTGD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.