missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MUC-1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£200.00 - £512.00
Specifications
| Antigen | MUC1 |
|---|---|
| Clone | 2336D |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18730137
![]() |
R&D Systems
MAB629812-SP |
25 μg |
£200.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18790507
![]() |
R&D Systems
MAB629812-100 |
100 μg |
£512.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MUC1 Monoclonal antibody specifically detects MUC1 in Human samples. It is validated for ELISASpecifications
| MUC1 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEM, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
| Chinese hamster ovary cell line CHO-derived recombinant human MUC-1, Met1-Ser380, Accession # P15941 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2336D | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P15941 | |
| 4582 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title