missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MTGR1 (aa 540-603) Control Fragment Recombinant Protein

Product Code. 30193590
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30193590

Brand: Invitrogen™ RP105562

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Myeloid tumor related gene (MTGR1) is found deleted in acute myeloid leukemia. Overexpression of MTGR1 can stimulate AML1-MTG8 to induce GCSF- dependent proliferation of L-G cells and to interfere with AML1- dependent transcription. AML1/MTG8 functions as a complex with MTGR1 and that the complex might be important in promoting leukemogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43439
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9139
Name Human MTGR1 (aa 540-603) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A430091M07; C330013D05Rik; CBFA2/RUN x 1 partner transcriptional co-repressor 2; CBFA2/RUN x 1 translocation partner 2; CBFA2T2; CBFA2T2 identified gene homolog; Cbfa2t2h; core-binding factor runt domain alpha subunit 2 translocated to 2 homolog; core-binding factor, runt domain, alpha subunit 2, translocated to, 2 (human); core-binding factor, runt domain, alpha subunit 2, translocated to, 2 homolog; core-binding factor, runt domain, alpha subunit 2; translocated to, 2; DKFZp313F2116; EHT; ETO homolog on chromosome 20; ETO homologous on chromosome 20; MTG8-like protein; MTG8-related protein 1; MTGR1; myeloid translocation gene-related protein 1; myeloid translocation-related protein 1; p85; Protein CBFA2T2; RP5-1137F22.1; translocated to, 2; ZMYND3; ZMYND3 CBFA2T2
Common Name MTGR1
Gene Symbol CBFA2T2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HRLCGQNLHGQSPHGQGRPLLPVGRGSSARSADCSVPSPALDKTSATTSRSSTPASVTAIDTNG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.