missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MTG16 (aa 4-74) Control Fragment Recombinant Protein

Product Code. 30210929
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210929

Brand: Invitrogen™ RP105373

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (42%), Rat (42%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-66294 (PA5-66294, PA5-66808 (PA5-66808. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the myeloid translocation gene family which interact with DNA-bound transcription factors and recruit a range of corepressors to facilitate transcriptional repression. The t(16;21)(q24;q22) translocation is one of the less common karyotypic abnormalities in acute myeloid leukemia. The translocation produces a chimeric gene made up of the 5'-region of the runt-related transcription factor 1 gene fused to the 3'-region of this gene. This gene is also a putative breast tumor suppressor. Alternative splicing results in transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75081
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 863
Name Human MTG16 (aa 4-74) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A630044F12Rik; AI465270; AW229127; CBFA2/RUN x 1 partner transcriptional co-repressor 3; CBFA2/RUN x 1 translocation partner 3; CBFA2T3; Cbfa2t3h; core-binding factor, runt domain, alpha subunit 2, translocated to, 3; core-binding factor, runt domain, alpha subunit 2, translocated to, 3 (human); core-binding factor, runt domain, alpha subunit 2; translocated to, 3; eight twenty one protein 2; ETO/MTG8-related protein ETO-2; Eto2; ETO-2; hMTG16; MTG16; MTG16HEL; MTG8-related gene 2; MTG8-related protein 2; MTGR2; myeloid translocation gene 8 and 16 b; myeloid translocation gene on chromosome 16 protein; myeloid translocation gene-related protein 2; protein CBFA2T3; Protein ETO-2; RUN x 1T3; translocated to, 3; zinc finger MYND domain-containing protein 4; ZMYND4
Common Name MTG16
Gene Symbol CBFA2T3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SRLRDRAASSASGSTCGSMSQTHPVLESGLLASAGCSAPRGPRKGGPAPVDRKAKASAMPDSPAEVKTQPR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.