missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MT-ND4L (aa 77-97) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67392 (PA5-67392. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
NADH:ubiquinone oxidoreductase (complex I) is an extremely complicated multiprotein complex located in the inner mitochondrial membrane. Human complex I is important for energy metabolism because its main function is to transport electrons from NADH to ubiquinone, which is accompanied by translocation of protons from the mitochondrial matrix to the intermembrane space. Human complex I appears to consist of 41 subunits. A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 4L (ND4L) is most likely a component of the hydrophobic protein fragment of Complex I. Mutations in the gene encodiing for ND4 are implicated in Leber hereditary optic neuropathy, a rare condition that can cause loss of central vision.
Specifications
Specifications
| Accession Number | P03901 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 4539 |
| Name | Human MT-ND4L (aa 77-97) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | mitochondrially encoded NADH 4 L dehydrogenase; MTND4L; mt-Nd4l; NADH dehydrogenase subunit 4 L; NADH4L; NADH-ubiquinone oxidoreductase chain 4 L; Nd4l |
| Common Name | MT-ND4L |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction