missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MSX1 (aa 115-143) Control Fragment Recombinant Protein

Código de producto. 30194107
Click to view available options
Quantity:
100 μL
Tamaño de la unidad:
100µL
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30194107

Marca: Invitrogen™ RP107940

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MSX1 is a member of the muscle segment homeobox gene family. It functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins. MSX1 may also have roles in limb-pattern formation, craniofacial development, particularly odontogenesis, and tumor growth inhibition. Mutations in the gene, which was once known as homeobox 7, have been associated with nonsyndromic cleft lip with or without cleft palate 5, Witkop syndrome, Wolf-Hirschom syndrome, and autosomoal dominant hypodontia.
TRUSTED_SUSTAINABILITY

Especificaciones

Accession Number P28360
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4487
Name Human MSX1 (aa 115-143) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA675338; AI324650; ECTD3; homeo box, msh-like 1; homeobox 7; homeobox protein Hox-7; Homeobox protein MSX-1; homeobox, msh-like 1; Hox7; Hox-7; Hox7.1; hox-7.1; HYD1; msh; msh homeo box 1; msh homeobox 1; msh homeobox 1-like protein; msh homeobox homolog 1; Msx1; muscle-segment homeobox; STHAG1
Common Name MSX1
Gene Symbol MSX1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LGHFSVGGLLKLPEDALVKAESPEKPERT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado