missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MSP/MST1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF352-SP
This item is not returnable.
View return policy
Description
MSP/MST1 Polyclonal specifically detects MSP/MST1 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| MSP/MST1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| D3F15S2, EC 3.4.21, hepatocyte growth factor-like protein homolog, HGFL, HGFLhepatocyte growth factor-like protein, macrophage stimulating 1 (hepatocyte growth factor-like), Macrophage stimulatory protein, Macrophage-stimulating protein, MSPDNF15S2, MST1, NF15S2, SF2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4485 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.05-0.2 ug/mL | |
| AAA59872 | |
| MST1 | |
| Mouse myeloma cell line NS0-derived recombinant human MSP/MST1 Gln19-Gly711 Accession # AAA59872 | |
| 25 μg | |
| Primary | |
| Detects human MSP/MST1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human HGF is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction