missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MSK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB2518-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MSK1/RPS6KA5 Monoclonal specifically detects MSK1/RPS6KA5 in Human samples. It is validated for Immunocytochemistry.
Especificaciones
| MSK1/RPS6KA5 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| O75582 | |
| RPS6KA5 | |
| E. coli-derived recombinant human MSK1 Met1-Ala802 Accession # O75582 | |
| 25 μg | |
| Primary | |
| Detects human MSK1 but not human MSK2 in direct ELISAs. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 252608 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11, MGC1911,90 kDa ribosomal protein S6 kinase 5, MSK1EC 2.7.11.1, MSPK1, Nuclear mitogen- and stress-activated protein kinase 1, ribosomal protein S6 kinase alpha-5, ribosomal protein S6 kinase, 90kD, polypeptide 5, ribosomal protein S6 kinase, 90kDa, polypeptide 5, RLPK, RPS6KA5, RSKL, RSK-like protein kinase, S6K-alpha-5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9252 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido