missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MSI1 (aa 217-274) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Musashi-1 belongs to the Musashi family of RNA-binding proteins which play a role in cell fate determination, and maintenance of stem-cell state. Musashi family proteins are also involved in post transcriptional gene regulation. The gene encodes a protein with two conserved tandem RNA recognition motifs with highly conserved RNP (ribonucleoprotein) motifs. In mammals, Musashi-1 is expressed in fetal kidney, brain, liver and lung, and in adult brain and pancreas. In humans, the gene is located on chromosome 12.
Specifications
Specifications
| Accession Number | O43347 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 4440 |
| Name | Human MSI1 (aa 217-274) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | fb40b12; hypothetical protein LOC541389; m-Msi-1; ms1; msi1; msi1b; Msi1h; Musahi1; musashi homolog 1; Musashi homolog 1(Drosophila); musashi RNA binding protein 1; musashi RNA binding protein 1 b; musashi RNA-binding protein 1; musashi1; musashi-1; RNA-binding protein Musashi homolog 1; RNA-binding protein Musashi homolog 1 L+35 bp; RNA-binding protein Musashi homolog 1 Long; RNA-binding protein Musashi homolog 1 b; wu:fb40b12; zgc:103751; zMsi1 |
| Common Name | MSI1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction