missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MS4A7 (aa 101-184) Control Fragment Recombinant Protein

Product Code. 30202334
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202334

Brand: Invitrogen™ RP90412

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (49%), Rat (49%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53603 (PA5-53603. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the membrane-spanning 4A gene family, members of which are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns in hematopoietic cells and nonlymphoid tissues. This family member is associated with mature cellular function in the monocytic lineage, and it may be a component of a receptor complex involved in signal transduction. This gene is localized to 11q12, in a cluster of other family members. At least four alternatively spliced transcript variants encoding two distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9GZW8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 58475
Name Human MS4A7 (aa 101-184) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4 SPAN2; CD20 antigen-like 4; CD20/Fc-epsilon-RI-beta family member 4; CD20L4; CFFM4; Four-span transmembrane protein 2; high affinity immunoglobulin epsilon receptor beta subunit; membrane spanning 4-domains A7; membrane-spanning 4-domains subfamily A member 7; membrane-spanning 4-domains subfamily A member 7; LOW QUALITY PROTEIN: membrane-spanning 4-domains subfamily A member 7; membrane-spanning 4-domains, subfamily A, member 7; MGC22368; MS4A7; MS4A8
Common Name MS4A7
Gene Symbol MS4A7
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LSIISGKQSTKPFDLSSLTSNAVSSVTAGAGLFLLADSMVALRTASQHCGSEMDYLSSLPYSEYYYPIYEIKDCLLTSVSLTGV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.