missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MS4A4A Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7797N-100UG
This item is not returnable.
View return policy
Description
MS4A4A Monoclonal specifically detects MS4A4A in Human samples. It is validated for Flow Cytometry.
Specifications
| MS4A4A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q96JQ5 | |
| MS4A4A | |
| Mouse myeloma cell line NS0 transfected with human MS4A4A Accession # Q96JQ5 | |
| RUO | |
| 51338 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 818112 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| 4SPAN1, CD20 antigen-like 1, CD20L1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4, MS4A4MGC22311, MS4A7 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human MS4A4A. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction