missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MRPL24 (aa 142-215) Control Fragment Recombinant Protein

Product Code. 30196389
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196389

Brand: Invitrogen™ RP108401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84193 (PA5-84193. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein which is more than twice the size of its E. coli counterpart. Sequence analysis identified two transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96A35
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79590
Name Human MRPL24 (aa 142-215) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2010005E08Rik; 2810470K06Rik; 39 S ribosomal protein L24, mitochondrial; 6720473G22Rik; AA407670; FLJ20917; hypothetical protein LOC532203; L24mt; MGC22737; MGC9831; Mitochondrial large ribosomal subunit protein uL24m; mitochondrial ribosomal protein L24; MRP-L18; MRPL24; MRP-L24; MRPL24 protein
Common Name MRPL24
Gene Symbol mrpl24
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ERVRVSTRSGRIIPKPEFPRADGIVPETWIDGPKDTSVEDALERTYVPCLKTLQEEVMEAMGIKETRKYKKVYW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.