missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human MRPL20 (aa 75-149) Control Fragment Recombinant Protein

Artikelnummer. 30195952
Klik for at se tilgængelige muligheder
Quantity:
100 μL
Pakningsstørrelse:
100µL
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 30195952

Brand: Invitrogen™ RP100655

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61417 (PA5-61417. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 21q. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Tekniske data

Accession Number Q9BYC9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55052
Name Human MRPL20 (aa 75-149) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2610008D01Rik; 39 S ribosomal protein L20, mitochondrial; 4930425I20Rik; L20mt; Mitochondrial large ribosomal subunit protein bL20m; mitochondrial ribosomal protein L20; MRPL20; MRP-L20
Common Name MRPL20
Gene Symbol MRPL20
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ASQEHGLKYPALIGNLVKCQVELNRKVLADLAIYEPKTFKSLAALASRRRHEGFAAALGDGKEPEGIFSRVVQYH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.