missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MRCK gamma (aa 1121-1197) Control Fragment Recombinant Protein

Product Code. 30212536
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212536

Brand: Invitrogen™ RP103424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63988 (PA5-63988. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CDC42 BPG serine/threonine kinase is encoded by the CDC42BPG gene. It acts as a downstream effector of CDC42 in cytoskeletal reorganization, and contributes to the actomyosin contractility required for cell invasion, through the regulation of MYPT1 and MLC2 phosphorylation. It belongs to the AGC protein kinase family, DMPK subfamily. It contains an N-terminal kinase domain, followed by a coiled-coil domain, a second coiled-coil domain containing a kinase inhibitory motif (KIM), a variable region, a cysteine-rich domain, a pleckstrin (PLEK; 173570) homology (PH) domain, a citron (605629) homology (CNH) domain, and a C-terminal CDC42 (116952)/RAC (602048) interactive binding (CRIB) domain.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6DT37
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55561
Name Human MRCK gamma (aa 1121-1197) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BC046418; CDC42 binding protein kinase gamma; CDC42 binding protein kinase gamma (DMPK-like); CDC42-binding protein kinase gamma; CDC42BPG; DMPK2; DMPK-like gamma; HSMDPKIN; KAPPA-200; MRCK gamma; MRCKG; MRCKgamma; Myotonic dystrophy kinase-related CDC42-binding kinase gamma; myotonic dystrophy protein kinase like protein; myotonic dystrophy protein kinase-like alpha; myotonic dystrophy protein kinase-like gamma; serine/threonine-protein kinase MRCK gamma
Common Name MRCK gamma
Gene Symbol CDC42BPG
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SNDIFQVGECRRVQQLTLSPSAGLLVVLCGRGPSVRLFALAELENIEVAGAKIPESRGCQVLAAGSILQARTPVLCV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.