missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MPG (aa 165-282) Control Fragment Recombinant Protein

Product Code. 30213446
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30213446

Brand: Invitrogen™ RP89471

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82252 (PA5-82252. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MPG functions in the hydrolysis of the deoxyribose N-glycosidic bond to excise 3-methyladenine, and 7-methylguanine from the damaged DNA polymer formed by alkylation lesions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29372
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4350
Name Human MPG (aa 165-282) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 3' end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on 16 p; 3-alkyladenine DNA glycosylase; 3-methyladenine DNA glycosidase; 3-methyladenine DNA glycosylase; 9830006D05; AAG; ADPG; AI326268; alkylpurine-DNA-N-glycosylase; ANPG; APNG; CRA36.1; CRA36.1 (3-methyl-adenine DNA glycosylase); DNA-3-methyladenine glycosylase; MDG; Mid1; MPG; N-methylpurine DNA glycosylase; N-methylpurine-DNA glycocylase; N-methylpurine-DNA glycosylase; N-methylpurine-DNA glycosylase, MPG; PIG11; PIG16; proliferation-inducing protein 11; proliferation-inducing protein 16
Common Name MPG
Gene Symbol MPG
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YFCMNISSQGDGACVLLRALEPLEGLETMRQLRSTLRKGTASRVLKDRELCSGPSKLCQALAINKSFDQRDLAQDEAVWLERGPLEPSEPAVVAAARVGVGHAGEWARKPLRFYVRGS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.