missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse ZAG Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £406.00
Specifications
| Antigen | ZAG |
|---|---|
| Clone | 842014 |
| Dilution | Immunocytochemistry 5-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18709377
![]() |
R&D Systems
MAB47641-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18723929
![]() |
R&D Systems
MAB47641-100 |
100 μg |
£406.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZAG Monoclonal specifically detects ZAG in Human, Mouse samples. It is validated for Immunocytochemistry.Specifications
| ZAG | |
| Immunocytochemistry 5-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Mouse | |
| P25311 | |
| 563 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 842014 | |
| Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| alpha-2-glycoprotein 1, zinc, alpha-2-glycoprotein 1, zinc-binding, AZGP1, ZA2G, ZAGzn-alpha-2-GP, zinc, zinc-alpha-2-glycoprotein, zn-alpha-2-glycoprotein, ZNGP1 | |
| Mouse myeloma cell line NS0-derived recombinant human ZAG Met1-Ser298 Accession # P25311 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human ZAG in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title