missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse XIAP Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB822-SP
This item is not returnable.
View return policy
Description
XIAP Monoclonal specifically detects XIAP in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.
Specifications
| XIAP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Knockout Validated | |
| P98170 | |
| XIAP | |
| E. coli-derived recombinant human XIAP Met1-Ser497 Accession # P98170 | |
| 25 μg | |
| Primary | |
| Detects human and mouse XIAP in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) cIAP-1 or rhcIAP-2 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot, KnockDown | |
| 117320 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| API3X-linked inhibitor of apoptosis protein, apoptosis inhibitor 3, baculoviral IAP repeat-containing 4, baculoviral IAP repeat-containing protein 4, BIRC4, BIRC4XLP2, E3 ubiquitin-protein ligase XIAP, EC 6.3.2.-, hIAP3, hIAP-3, hILP, IAP3, IAP-3, IAP-like protein, ILP, ILP1, Inhibitor of apoptosis protein 3, MIHA, X-linked IAP, X-linked inhibitor of apoptosis | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 331 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction