missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Wnt-9b Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3669
This item is not returnable.
View return policy
Description
Wnt-9b Monoclonal specifically detects Wnt-9b in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Wnt-9b | |
| Monoclonal | |
| LYOPH | |
| Western Blot 5 ug/mL | |
| O35468.2 | |
| WNT9B | |
| E. coli-derived recombinant mouse Wnt-9b Ala46-Glu90 & Lys228-Tyr298 Accession # O35468.2 | |
| 100 μg | |
| Primary | |
| Detects human Wnt-9b in Western blots. Detects recombinant mouse Wnt-9b in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Wnt-2, -6, -7a, -7b, -9a, recombinant mouse Wnt-1, -2b, -4, -5a, -5b, -8a, -8b, -10a, -10b, -11, or -16 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 406612 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Protein Wnt-14b, wingless-type MMTV integration site family, member 15, wingless-type MMTV integration site family, member 9B, Wnt-14b, WNT14Bprotein Wnt-9b, Wnt-15, WNT15Protein Wnt-15, Wnt9b | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7484 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction