missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Wnt-5a Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 13 publications
Brand: R&D Systems MAB645
This item is not returnable.
View return policy
Description
Wnt-5a Monoclonal specifically detects Wnt-5a in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Wnt-5a | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P22725 | |
| WNT5A | |
| CHO-derived recombinant mouse Wnt-5a Gln38-Lys380 Accession # P22725 | |
| 100 μg | |
| Primary | |
| Detects mouse Wnt-5a in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Wnt-1, 2b, 3a, 4, 5b, 8a, 8b, 9b, 10a, 10b, 11, or 16 is observed. | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry | |
| 442625 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, Wnt5a, WNT-5A protein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7474 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction