missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Wnt-4 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
£234.34 - £1406.01
Specifications
Antigen | Wnt-4 |
---|---|
Clone | 55010 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18142461
|
R&D Systems
MAB475-050 |
50 μg |
£234.34
50µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18120541
|
R&D Systems
MAB475-500 |
500 μg |
£1406.01
500µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Wnt-4 Monoclonal specifically detects Wnt-4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Wnt-4 | |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rat | |
Mouse | |
P22724 | |
54361 | |
E. coli-derived recombinant mouse Wnt-4 aa 37-76 and 222-295 Accession # P22724 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Wnt-4 in direct ELISAs and Western blots. |
55010 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
protein Wnt-4, SERKAL, wingless-type MMTV integration site family, member 4, Wnt4, WNT-4 | |
WNT4 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |