missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse VGF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6317-SP
This item is not returnable.
View return policy
Description
VGF Monoclonal specifically detects VGF in Human, Mouse samples. It is validated for Immunohistochemistry.
Specifications
| VGF | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| O15240 | |
| VGF | |
| E. coli-derived recombinant human VGF Thr554-Pro615 Accession # O15240 | |
| 25 μg | |
| Primary | |
| Detects human and mouse VGF in direct ELISAs. | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry | |
| 619707 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| neuro-endocrine specific protein VGF, neurosecretory protein VGF, VGF nerve growth factor inducible | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7425 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction