Learn More
Human/Mouse VGF Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB6317
 View more versions of this product
Product Code. 18111671
  • £370.00 / 100µg
Description
VGF Monoclonal specifically detects VGF in Human, Mouse samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VGF
Monoclonal
LYOPH
Immunohistochemistry 8-25 ug/mL
O15240
VGF
E. coli-derived recombinant human VGF Thr554-Pro615 Accession # O15240
100 μg
Primary
Detects human and mouse VGF in direct ELISAs.
Human, Mouse
Purified
Immunohistochemistry
619707
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
neuro-endocrine specific protein VGF, neurosecretory protein VGF, VGF nerve growth factor inducible
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
7425
Sterile PBS to a final concentration of 0.5 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG3