missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse VEGF-B167/186 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£155.00 - £1445.00
Specifications
| Antigen | VEGF-B |
|---|---|
| Clone | 58013 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
Description
VEGF-B Monoclonal specifically detects VEGF-B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| VEGF-B | |
| Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Mouse | |
| AAB06274 | |
| 7423 | |
| E. coli-derived recombinant human VEGF-B 167 Pro22-Arg188 Accession # AAB06274 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and mouse VEGF-B in direct ELISAs and Western blots. Recognizes both the VEGF-B 167 and VEGF-B 186 isoforms. In Western blots, no cross-reactivity with recombinant human (rh) VEGF, rhVEGF-C, rhVEGF-D, rhCTGF, rhPlGF, rhLDGF, or rhPDGF is observed. |
| 58013 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL | |
| VEGFB | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title