missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Vav-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB3786
This item is not returnable.
View return policy
Description
Vav-1 Monoclonal specifically detects Vav-1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Vav-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P15498 | |
| VAV1 | |
| E. coli-derived recombinant human Vav-1 Trp25-Leu228 Accession # P15498 | |
| 100 μg | |
| Primary | |
| Detects human and mouse Vav-1 in Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 591529 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| oncogene vav, proto-oncogene vav, vav 1 guanine nucleotide exchange factor, vav proto-oncogene, Vav1, vav-T, VAVvav 1 oncogene | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7409 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu