missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse TREM2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 13 publications
Brand: R&D Systems MAB17291-100
This item is not returnable.
View return policy
Description
TREM2 Monoclonal specifically detects TREM2 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry, CyTOF-ready, Knockout Validated.
Specifications
| TREM2 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, ELISA, Immunohistochemistry 0.5-15 ug/mL, Immunocytochemistry 5-25 ug/mL, CyTOF-ready, Knockout Validated | |
| Q99NH8 | |
| TREM2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TREM-2 extracellular domain Accession # Q99NH8 | |
| 100 μg | |
| Primary | |
| Detects human and mouse TREM-2 in direct ELISAs and Western blots. Stains TREM-2 transfectants but not TREM-1 transfectants. | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, CyTOF, Immunoassay | |
| 237920 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54209 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction