missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Mouse Total HIF-1 alpha/HIF1A DuoSet IC ELISA

Product Code. 30021563
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30021563

Brand: R&D Systems™ DYC1935E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human/Mouse Total HIF-1 alpha DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human/Mouse Total HIF-1 alpha DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse HIF-1 alpha.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Pk. (for 15 Plates)
Species Human, Mouse
Conjugate Biotin
Assay Range 125.00 to 8000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 3091
Synonym AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF 1A, HIF1, HIF1 alpha, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8
Packaging Quantity 15 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.