missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Tmp21/p23 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7630-SP
This item is not returnable.
View return policy
Description
Tmp21/p23 Polyclonal specifically detects Tmp21/p23 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.
Specifications
| Tmp21/p23 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 21 kDa transmembrane-trafficking protein, p23, P24(DELTA), p24delta, S31I125, S31III125, TMED10, TMP21Tmp-21-I, transmembrane emp24 domain-containing protein 10, transmembrane emp24-like trafficking protein 10 (yeast), Transmembrane protein Tmp21,21 kDa transmembrane trafficking protein | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10972 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoassay | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 1-15 ug/mL, Knockout Validated | |
| P49755 | |
| TMED10 | |
| E. coli-derived recombinant human Tmp21/p23 Ile32-Arg185 Accession # P49755 | |
| 25 μg | |
| Primary | |
| Detects human and mouse in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction