missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse TLR9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£170.00 - £369.00
Specifications
| Antigen | TLR9 |
|---|---|
| Clone | 980915 |
| Dilution | Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 3-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18737848
![]() |
R&D Systems
MAB36581-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18760788
![]() |
R&D Systems
MAB36581-100 |
100 μg |
£369.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR9 Monoclonal specifically detects TLR9 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
| TLR9 | |
| Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human, Mouse | |
| Q9NR96 | |
| 54106 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 980915 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| Human TLR9 synthetic peptide Accession # Q9NR96 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TLR9 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title