missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse TIMP-4 Antibody, R&D Systems™

Goat Polyclonal Antibody
£168.00 - £509.00
Specifications
| Antigen | TIMP-4 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18737287
![]() |
R&D Systems
AF974-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18060302
![]() |
R&D Systems
AF974 |
100 μg |
£509.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMP-4 Polyclonal specifically detects TIMP-4 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
| TIMP-4 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human, Mouse | |
| metalloproteinase inhibitor 4, TIMP metallopeptidase inhibitor 4, TIMP4, TIMP-4, tissue inhibitor of metalloproteinase 4, Tissue inhibitor of metalloproteinases 4 | |
| TIMP4 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q99727 | |
| 7079 | |
| Mouse myeloma cell line NS0-derived recombinant human TIMP-4 Cys30-Pro224 Accession # Q99727 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human and mouse TIMP-4 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) TIMP-1, rhTIMP-2, and rhTIMP-3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title