missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Stathmin-2/STMN2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6930
This item is not returnable.
View return policy
Description
Stathmin-2/STMN2 Monoclonal specifically detects Stathmin-2/STMN2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Stathmin-2/STMN2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.05 - 0.2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q93045 | |
| STMN2 | |
| E. coli-derived recombinant human Stathmin-2/STMN2 Tyr26-Gly179 Accession # Q93045 | |
| 100 μg | |
| Primary | |
| Detects human Stathmin-2/STMN2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Stathmin-1, -3, or -4 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 684433 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ34868, FLJ50995, neuron-specific growth-associated protein, Protein SCG10, SCG10, SCG10neuronal growth-associated protein (silencer element), SCGN10, SCGN10stathmin-2, Stathmin2, stathmin-like 2, STMN2, superior cervical ganglia, neural specific 10, Superior cervical ganglion-10 protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11075 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction