missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse SSEA-4 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 14 publications
Brand: R&D Systems FAB1435P-100
This item is not returnable.
View return policy
Description
SSEA-4 Monoclonal specifically detects SSEA-4 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| SSEA-4 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 2120Ep human embryonal carcinoma cell line | |
| 100 Tests | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| MC-813-70 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Recognizes a carbohydrate epitope of SSEA-4 (1, 2). | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction