missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse SSEA-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 17 publications
Brand: R&D Systems MAB2155
This item is not returnable.
View return policy
Description
SSEA-1 Monoclonal specifically detects SSEA-1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunocytochemistry, CyTOF-reported.
Specifications
| SSEA-1 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 μg/106 cells, Immunoprecipitation, Immunocytochemistry 8 to 25 μg/mL, CyTOF-reported | |
| alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA1, SSEA-1, stage-specific embryonic antigen 1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 2526 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Flow Cytometry, Immunoprecipitation, Immunocytochemistry, CyTOF | |
| MC-480 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FUT4 | |
| F9 mouse teratocarcinoma stem cells | |
| 100 μg | |
| Primary | |
| Detects human and mouse SSEA-1. | |
| Human, Mouse | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction