missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse SorCS3 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB3067
This item is not returnable.
View return policy
Description
SorCS3 Monoclonal specifically detects SorCS3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| SorCS3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8VI51 | |
| SORCS3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse SorCS3 Ala131-Ser1122 Accession # Q8VI51 | |
| 100 μg | |
| Primary | |
| Detects human and mouse SorCS3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SorCS1 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 339624 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 6330404A12Rik, KIAA1059SORCS, sortilin-related VPS10 domain containing receptor 3, VPS10 domain receptor protein SORCS 3 (SORCS3), VPS10 domain-containing receptor SorCS3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 22986 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction