missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Sonic Hedgehog/Shh N-Terminus Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 13 publications
Brand: R&D Systems AF464
This item is not returnable.
View return policy
Description
Sonic Hedgehog/Shh Polyclonal specifically detects Sonic Hedgehog/Shh in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Sonic Hedgehog/Shh | |
| Polyclonal | |
| Unconjugated | |
| Q62226 | |
| SHH | |
| E. coli-derived recombinant mouse Sonic Hedgehog/Shh N-Terminus Cys25-Gly198 (Lys122Arg) Accession # Q62226 | |
| 100 μg | |
| Primary | |
| Detects human and mouse Sonic Hedgehog/Shh N-Terminus in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant mouse (rm) Dhh N-Terminus and rmIhh N-Terminus is observed. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6469 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction