missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse SOCS-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB5696
This item is not returnable.
View return policy
Description
SOCS-3 Monoclonal specifically detects SOCS-3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| SOCS-3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| O14543 | |
| SOCS3 | |
| E. coli-derived recombinant human SOCS-3 Met1-Leu225 Accession # O14543 | |
| 100 μg | |
| Primary | |
| Detects human or mouse SOCS-3 in Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 516919 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ATOD4, CIS3, CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9021 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction