missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Serpin I1 Antibody, R&D Systems™

Rat Monoclonal Antibody
£157.00 - £337.00
Specifications
| Antigen | Serpin I1/Neuroserpin |
|---|---|
| Clone | 437016 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18734428
![]() |
R&D Systems
MAB3124-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18181891
|
R&D Systems
MAB3124 |
100 μg |
£337.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin I1/Neuroserpin Monoclonal specifically detects Serpin I1/Neuroserpin in Human, Mouse samples. It is validated for Western Blot.Specifications
| Serpin I1/Neuroserpin | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human, Mouse | |
| AAH18043 | |
| 5274 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin I1 Ala19-Leu410 Accession # AAH18043 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and mouse SerpinI1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross reactivity with recombinant mouse Serpin I1 is observed and no cross-reactivity with recombinant human Serpin A1, A3, A4, A5, A12, B6, B8, B9, C1, D1, F2, G1, or I2 is observed. |
| 437016 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp781N13156, Neuroserpin, Peptidase inhibitor 12, PI12, PI-12, PI12Serpin I1, serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 1, serpin peptidase inhibitor, clade I (neuroserpin), member 1 | |
| SERPINI1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title