missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Semaphorin 6B Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF2094
Additional Details : Weight : 0.00970kg
Description
Semaphorin 6B Polyclonal specifically detects Semaphorin 6B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Semaphorin 6B | |
Polyclonal | |
Unconjugated | |
Q9H3T3 | |
SEMA6B | |
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6B Leu26-Ser603 Accession # Q9H3T3 | |
100 μg | |
Primary | |
Detects human and mouse Semaphorin 6B in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) Semaphorin 6C is observed, 5% cross-reactivity with rhSemaphorin 6D is observed and less than 1% cross-reactivity with rhSemaphorin 3A, rhSemaphorin 3B, rhSemaphorin 6A, rhSemaphorin 7A, recombinant mouse (rm) Semaphorin 3C, rmSemaphorin 3E, and rmSemaphorin 3F is observed. | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B, Sema6B, SEMAN, semaphorin VIB, semaphorin Z, semaphorin-6B, semaphorin-6Ba, SEMA-VIB, SEMAZ, SEM-SEMA-Y, UNQ1907/PRO4353 | |
Goat | |
Affinity Purified | |
RUO | |
10501 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only