missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Semaphorin 3E Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF3239
This item is not returnable.
View return policy
Description
Semaphorin 3E Polyclonal specifically detects Semaphorin 3E in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| Semaphorin 3E | |
| Polyclonal | |
| Unconjugated | |
| O15041 | |
| SEMA3E | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 3E Thr25-Ser775 (Arg557Ala and Arg560Ala) Accession # O15041 | |
| 100 μg | |
| Primary | |
| Detects human Semaphorin 3E in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) Semaphorin 3A, rhSemaphorin 3C, rhSemaphorin 3F, and rhSemaphorin 3G is observed. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| (semaphorin) 3E, coll-5, KIAA0331M-SEMAH, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema3E, SEMAH, SEMAHM-SemaK, semaphorin-3E | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9723 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction