missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse SDPR Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF5759
This item is not returnable.
View return policy
Description
SDPR Polyclonal specifically detects SDPR in Human, Mouse samples. It is validated for Western Blot.
Specifications
| SDPR | |
| Polyclonal | |
| Unconjugated | |
| O95810 | |
| SDPR | |
| E. coli-derived recombinant human SDPR Gly2-Val180 Accession # O95810 | |
| 100 μg | |
| Primary | |
| Detects human and mouse SDPR in Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| cavin-2, Phosphatidylserine-binding protein, PS-p68, PS-p68serum deprivation response (phosphatidylserine-binding protein), SDR, SDRphosphatidylserine binding protein, serum deprivation response, serum deprivation response (phosphatidylserine binding protein), serum deprivation-response protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8436 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction