missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse S100A4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4138
This item is not returnable.
View return policy
Description
S100A4 Polyclonal specifically detects S100A4 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Knockout Validated.
Specifications
| S100A4 | |
| Polyclonal | |
| Unconjugated | |
| P07091 | |
| S100A4 | |
| E. coli-derived recombinant mouse S100A4 Ala2-Lys101 Accession # P07091 | |
| 100 μg | |
| Primary | |
| Detects human and mouse S100A4 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) S100A8, rmS100A6, rmS100A9, rmS100A10, rmS100A13, rmS100A16, recombinant human (rh) S100A1, rhS100A7, rhS100A11, rhS100B, and rhS100P is observed. | |
| Human, Mouse | |
| IgG |
| Western Blot, KnockDown | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Knockout Validated | |
| 18A2, 42A, Calvasculin, CAPL, CAPLS100 calcium binding protein A4 (calcium protein, calvasculin, metastasin, fibroblast-specific protein-1,42A, FSP1, leukemia multidrug resistance associated protein, malignant transformation suppression 1, Metastasin, MTS1, murine placental homolog), P9KA, PEL98, Placental calcium-binding protein, Protein Mts1, protein S100-A4, S100 calcium binding protein A4, S100 calcium-binding protein A4, S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog) | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6275 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction