missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse ROR gamma t/RORC2/NR1F3 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems IC6006A-100
This item is not returnable.
View return policy
Description
ROR gamma t/RORC2/NR1F3 Monoclonal specifically detects ROR gamma t/RORC2/NR1F3 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| ROR gamma t/RORC2/NR1F3 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| RORC | |
| Human ROR gamma t peptide Met1-Arg10 Accession # P51449 | |
| 100 Tests | |
| Primary | |
| Detects human and mouse ROR gamma t/RORC2 in direct ELISAs. In direct ELISAs, no cross-reactivity with human and mouse RORC. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 600380 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| P51449 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6097 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction