missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse RIPK4/RIP4 Antibody, R&D Systems™

Goat Polyclonal Antibody
£218.00 - £638.00
Specifications
| Antigen | RIPK4 |
|---|---|
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 0.3-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18768106
![]() |
R&D Systems
AF10002-SP |
25 μg |
£218.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18796466
![]() |
R&D Systems
AF10002-100 |
100 μg |
£638.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RIPK4 Polyclonal antibody specifically detects RIPK4 in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| RIPK4 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Goat | |
| Human, Mouse | |
| P57078 | |
| 54101 | |
| IgG | |
| Antigen Affinity-purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Western Blot 2 ug/mL, Immunohistochemistry 0.3-15 ug/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ANKK2, ANKRD3, ANKRD3MGC129993, ankyrin repeat domain 3, Ankyrin repeat domain-containing protein 3, DIKMGC129992, EC 2.7.11, EC 2.7.11.1, PKC-delta-interacting protein kinase, PKK, receptor-interacting serine/threonine-protein kinase 4, receptor-interacting serine-threonine kinase 4, RIP4, serine/threonine-protein kinase ANKRD3 | |
| E. coli-derived recombinant human/mouse RIPK4/RIP4, Thr218-Thr450, Accession # P57078 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title