missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse RIAM/APBB1IP Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7279-SP
This item is not returnable.
View return policy
Description
RIAM/APBB1IP Polyclonal specifically detects RIAM/APBB1IP in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| RIAM/APBB1IP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| amyloid beta (A4) precursor protein-binding, family B, member 1 interactingprotein, amyloid beta A4 precursor protein-binding family B member 1-interacting protein, APBB1-interacting protein 1, APBB1IP, INAG1, PREL1, PREL-1, proline rich EVH1 ligand 1, Proline-rich EVH1 ligand 1, Proline-rich protein 73, Rap1-GTP-interacting adapter molecule, Rap1-interacting adaptor molecule, RARP1, Retinoic acid-responsive proline-rich protein 1, RIAMRARP-1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 54518 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q7Z5R6 | |
| APBB1IP | |
| E. coli-derived recombinant human RIAM/APBB1IP Gly2-Ser89 Accession # Q7Z5R6 | |
| 25 μg | |
| Primary | |
| Detects recombinant human RIAM/APBB1IP in direct ELISAs. Detects human and mouse RIAM/APBB1IP in Western blots | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction