missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse RGM-C/Hemojuvelin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF3720-SP
This item is not returnable.
View return policy
Description
RGM-C/Hemojuvelin Polyclonal specifically detects RGM-C/Hemojuvelin in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| RGM-C/Hemojuvelin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DL-M, haemojuvelin, hemochromatosis type 2 (juvenile), hemojuvelin, HFE2, HFE2AMGC23953, HJV, HJVHemochromatosis type 2 protein, JH, repulsive guidance molecule c, RGMC, RGMCRGM domain family member C | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 148738 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q6ZVN8 | |
| HFE2 | |
| Mouse myeloma cell line NS0-derived recombinant human RGM-C isoform a (R&D Systems, Catalog # 3720-RG) Gln36-Asp400 Accession # Q6ZVN8 | |
| 25 μg | |
| Primary | |
| Detects human and mouse RGM-C in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) RGM-B is observed and less than 5% cross-reactivity with rhRGM-A is observed. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction