Learn More
Human/Mouse/Rat XIAP Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 14 publications
Brand:  R&D Systems AF8221-SP
Product Code. 18745038
  • £187.00 / 25µg
Description
XIAP Polyclonal specifically detects XIAP in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
XIAP
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
API3X-linked inhibitor of apoptosis protein, apoptosis inhibitor 3, baculoviral IAP repeat-containing 4, baculoviral IAP repeat-containing protein 4, BIRC4, BIRC4XLP2, E3 ubiquitin-protein ligase XIAP, EC 6.3.2.-, hIAP3, hIAP-3, hILP, IAP3, IAP-3, IAP-like protein, ILP, ILP1, Inhibitor of apoptosis protein 3, MIHA, X-linked IAP, X-linked inhibitor of apoptosis
Goat
Affinity Purified
RUO
331
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Immunohistochemistry, KnockDown
LYOPH
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated
P98170
XIAP
E. coli-derived recombinant human XIAP Met1-Ser497 (Ser162Cys) Accession # P98170
25 μg
Primary
Detects human, mouse and rat XIAP in Western blots.
Human, Mouse, Rat