missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat WWOX Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6399
This item is not returnable.
View return policy
Description
WWOX Polyclonal specifically detects WWOX in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| WWOX | |
| Polyclonal | |
| Unconjugated | |
| Q9NZC7 | |
| WWOX | |
| E. coli-derived recombinant human WWOX Asn36-Val126 Accession # Q9NZC7 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat WWOX in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| D16S432E, EC 1.1.1, EC 1.1.1.-, FOR, FORHHCMA56, FRA16D, Fragile site FRA16D oxidoreductase, SDR41C1, short chain dehydrogenase/reductase family 41C, member 1, WOX1, WOX1PRO0128, WW domain containing oxidoreductase, WW domain-containing oxidoreductase, WW domain-containing protein WWOX | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 51741 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction