missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Vimentin Antibody, R&D Systems™

Mouse Monoclonal Antibody
£184.00 - £397.00
Specifications
| Antigen | Vimentin |
|---|---|
| Clone | 979517 |
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL, Knockout Validated 5-15 ug/mL, Dual RNAscope ISH-IHC 5-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoassay, In Situ Hybridization (ISH) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18704889
![]() |
R&D Systems
MAB21052-SP |
25 μg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18710788
![]() |
R&D Systems
MAB21052-100 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Knockout Validated, Dual RNAscope ISH-IHC.Specifications
| Vimentin | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL, Knockout Validated 5-15 ug/mL, Dual RNAscope ISH-IHC 5-25 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human, Mouse, Rat | |
| P08670 | |
| 7431 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 979517 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoassay, In Situ Hybridization (ISH) | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| FLJ36605, VIM, vimentin | |
| E.coli-derived recombinant human Vimentin Ser2-Glu466 Accession # P08670 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Vimentin in direct ELISAs. Detects human, mouse, and rat Vimentin in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title