missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat UBR5 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6404
This item is not returnable.
View return policy
Description
UBR5/EDD Polyclonal specifically detects UBR5/EDD in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| UBR5/EDD | |
| Polyclonal | |
| Unconjugated | |
| O95071 | |
| UBR5 | |
| E. coli-derived recombinant human UBR5 Met1-Asp227 Accession # O95071 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat UBR5 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| DD5, E3 ubiquitin protein ligase, HECT domain containing, 1, E3 ubiquitin-protein ligase, HECT domain-containing 1, EC 6.3.2, EC 6.3.2.-, EDD, EDD1, EDD1E3 ubiquitin-protein ligase UBR5, EDDE3 identified by differential display, hHYD, HYD, HYDFLJ11310, Hyperplastic discs protein homolog, KIAA0896MGC57263, progestin induced protein, Progestin-induced protein, ubiquitin protein ligase E3 component n-recognin 5, ubiquitin-protein ligase | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 51366 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction