missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat TrkB Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
£170.00 - £1795.00
Specifications
| Antigen | TrkB |
|---|---|
| Clone | 75133 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18720638
![]() |
R&D Systems
MAB397-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18784968
![]() |
R&D Systems
MAB397-100 |
100 μg |
£456.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18782337
![]() |
R&D Systems
MAB397-500 |
500 μg |
£1795.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TrkB Monoclonal specifically detects TrkB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| TrkB | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Mouse, Rat | |
| Q16620 | |
| 4915 | |
| Mouse myeloma cell line NS0-derived recombinant human TrkB Cys32-His430 Accession # Q16620 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TrkB in direct ELISAs and Western blots. In ELISAs and Western blots, no cross-reactivity with recombinant human (rh) TrkA, rhTrkC, or recombinant rat TrkA is observed. |
| 75133 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, NTRK2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B | |
| NTRK2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title