missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Total GSK-3 alpha/beta DuoSet IC, R&D Systems™
Brand: R&D Systems DYC2157-2
This item is not returnable.
View return policy
Description
Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.